Learn More
Grainger Edwards Signaling Tone Generator, 120V AC, Zinc Alloy, 1 ea
Catalog No. 502791844
Encompass_Preferred
This heavy-duty, tone-selectable industrial signaling device is designed for demanding environments requiring high-decibel output and microprocessor reliability. It delivers a sound output of up to 110 dB at 10 ft. (3.05 m) and can be activated via field-wired contacts or an external 120 V AC voltage source, such as a PLC output.
  • Features 27 distinctive tones selectable via internal dip switches
  • Includes a built-in cascading priority system
  • Constructed from corrosion-resistant zinc alloy with an IP44 rating
  • Designed for 1/2 in. conduit or surface mounting
  • Suitable for both indoor and outdoor applications
  • PLC compatible with 120 V AC input board
Catalog No. 50-279-1844 Supplier Grainger Supplier No. 16G822
This heavy-duty, tone-selectable industrial signaling device is designed for demanding environments requiring high-decibel output and microprocessor reliability. It delivers a sound output of up to 110 dB at 10 ft. (3.05 m) and can be activated via field-wired contacts or an external 120 V AC voltage source, such as a PLC output.
  • Features 27 distinctive tones selectable via internal dip switches
  • Includes a built-in cascading priority system
  • Constructed from corrosion-resistant zinc alloy with an IP44 rating
  • Designed for 1/2 in. conduit or surface mounting
  • Suitable for both indoor and outdoor applications
  • PLC compatible with 120 V AC input board