Learn More
Grainger Edwards Signaling Tone Generator, Zinc Alloy, 55 Tones, 120-240V AC/125-250V DC
Catalog No. 502791847
Encompass_Preferred
The Adaptatone Millennium is a high-output electronic signal device designed for demanding industrial environments. It provides reliable audible signaling for both indoor and outdoor applications, operating on 120 V to 240 V AC or 125 V to 250 V DC. Featuring microcomputer reliability, this unit allows for sophisticated tone management with a built-in cascading priority system.
  • Up to 55 user-selectable tones
  • Adjustable speaker direction and output level
  • Built-in cascading priority system for active tone inputs
  • UL and cUL listed for Class I, Division 2, Groups A, B, C and D; Class II, Division 2, Groups F and G; and Class III hazardous locations
  • NEMA 3R and IP44 enclosure ratings
  • Durable zinc alloy construction
  • Suitable for surface or 1/2 in. conduit mounting
Catalog No. 50-279-1847 Supplier Grainger Supplier No. 16G831
The Adaptatone Millennium is a high-output electronic signal device designed for demanding industrial environments. It provides reliable audible signaling for both indoor and outdoor applications, operating on 120 V to 240 V AC or 125 V to 250 V DC. Featuring microcomputer reliability, this unit allows for sophisticated tone management with a built-in cascading priority system.
  • Up to 55 user-selectable tones
  • Adjustable speaker direction and output level
  • Built-in cascading priority system for active tone inputs
  • UL and cUL listed for Class I, Division 2, Groups A, B, C and D; Class II, Division 2, Groups F and G; and Class III hazardous locations
  • NEMA 3R and IP44 enclosure ratings
  • Durable zinc alloy construction
  • Suitable for surface or 1/2 in. conduit mounting