Learn More
Abcam Multiple x IHC Detection kit (TSA amplification), 90 Tests
Catalog No. 502660375
Encompass_Preferred
Multiplex IHC Detection kit (TSA amplification) is used for multiplex immunohistochemistry (mIHC) with tyramide signal amplification (TSA) on frozen tissue sections, which results in 10-50 times signal amplification compared with directly conjugated antibodies.
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-266-0375 Supplier Abcam Supplier No. AB31282790TEST
Multiplex IHC Detection kit (TSA amplification) is used for multiplex immunohistochemistry (mIHC) with tyramide signal amplification (TSA) on frozen tissue sections, which results in 10-50 times signal amplification compared with directly conjugated antibodies.
The product is subject to the following: Abcam Restricted Use Statement