Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Microbiologics™ Multiplex Vaginal Verification Panel
SDP

Catalog No. 23047421 Shop All Microbiologics Products
Change view
Click to view available options
Quantity:
10 Swabs
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-047-421 10 Swabs
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 23-047-421 Supplier Microbiologics™ Supplier No. 8245
Add to Cart
Add to Cart
This item is not returnable. View return policy

Multiplex Vaginal Verification Panel contains two inactivated positive control 1 swabs, two inactivated positive control 2 swabs, two inactivated positive control 3 swabs, two inactivated positive control 4 swabs and two inactivated negative control swabs (10 swabs total).

  • Multiplex Vaginal Verification Panel contains two inactivated positive control 1 swabs, two inactivated positive control 2 swabs, two inactivated positive control 3 swabs, two inactivated positive control 4 swabs and two inactivated negative control swabs (10 swabs total).
  • Positive Control 1 (two swabs):
    • Atopobium vaginae
    • BVAB2 surrogate
  • Positive Control 2 (two swabs):
    • Candida albicans
  • Positive Control 3 (two swabs):
    • Candida glabrata
  • Positive Control 4 (two swabs):
    • Trichomonas vaginalis
  • Negative Control (two swabs):
    • Lactobacillus acidophilus

Specifications

ATCC® Proprietary
Biosafety Level 1
Certifications/Compliance ISO 17025, ISO 13485, ISO 17034
Detects Bacterial Vaginosis-Causing Pathogens
Manufacturing Origin United States
Product Type Inactivated Swab
Sterility Sterile
Target Organism Class Gram-positive Organisms
For Use With (Application) Quality Control
Quantity 10 Swabs
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.