Learn More
Grainger Multipurpose Grease Cartridge 14 oz
Catalog No. 502763793
Encompass_Preferred
Sta-Lube Multi-Purpose Grease is a lithium complex, NLGI Grade 2 grease formulated for multi-purpose applications. It provides excellent resistance to water washout and spray-off, has a dropping point over 500°F, and protects against rust and corrosion. This 14 oz cartridge can be applied using a standard grease gun for lubricating bearings, bushings, universal joints and chassis. It's recommended for use in industrial, fleet, marine, farm and automotive applications.
Catalog No. 50-276-3793 Supplier Grainger Supplier No. 16W690
Sta-Lube Multi-Purpose Grease is a lithium complex, NLGI Grade 2 grease formulated for multi-purpose applications. It provides excellent resistance to water washout and spray-off, has a dropping point over 500°F, and protects against rust and corrosion. This 14 oz cartridge can be applied using a standard grease gun for lubricating bearings, bushings, universal joints and chassis. It's recommended for use in industrial, fleet, marine, farm and automotive applications.