Learn More
Grainger Clauss Straight Trimmer, Stainless Steel, Adjustable Tension, 8 in., 1 ea.
Catalog No. 502782189
Encompass_Preferred
These 8 in. straight trimmers are designed for heavy-duty cutting tasks across a variety of materials, including cardboard, fabric, leather, plastics, rope, rubber, and vinyl. Featuring a durable stainless steel construction, these versatile tools provide a reliable cutting action for multi-purpose applications.
  • Stainless steel blades with adjustable screw-type pivot tension
  • Large, ergonomic glass-filled nylon handles in black
  • Enlarged lower bow designed to accommodate gloved hands
  • Oversized fulcrum for enhanced cutting power
  • Pointed tip with a 3 1/2 in. length of cut
  • Ambidextrous design for versatile use
Catalog No. 50-278-2189 Supplier Grainger Supplier No. 19T041
These 8 in. straight trimmers are designed for heavy-duty cutting tasks across a variety of materials, including cardboard, fabric, leather, plastics, rope, rubber, and vinyl. Featuring a durable stainless steel construction, these versatile tools provide a reliable cutting action for multi-purpose applications.
  • Stainless steel blades with adjustable screw-type pivot tension
  • Large, ergonomic glass-filled nylon handles in black
  • Enlarged lower bow designed to accommodate gloved hands
  • Oversized fulcrum for enhanced cutting power
  • Pointed tip with a 3 1/2 in. length of cut
  • Ambidextrous design for versatile use