Learn More
Electron Microscopy Sciences Multi-Purpose Sample Cups, Capacity: 0.5 mL. Dimensions: 13.55mm x 24.50mm., 1000/PK
SDP
Catalog No. 502090601
Encompass_Preferred
These multi-purpose non-sterile sample cups are available in a variety of sizes. Made polystyrene, can be used for many laboratory applications. They provide excellent optical properties and are made to precise specifications. Autoclavable with a temperature range of -10° to +70° celsius. Capacity: 0.5 mL. Dimensions: 13.55mm x 24.50mm .
Catalog No. 50-209-0601 Supplier Electron Microscopy Sciences Supplier No. 6099404
These multi-purpose non-sterile sample cups are available in a variety of sizes. Made polystyrene, can be used for many laboratory applications. They provide excellent optical properties and are made to precise specifications. Autoclavable with a temperature range of -10° to +70° celsius. Capacity: 0.5 mL. Dimensions: 13.55mm x 24.50mm .