Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Axygen™ MultiRack Bulk Tip System
GREENER_CHOICE

Catalog No. 07202390 Shop All Corning Products
Change view
Click to view available options
Volume (Metric):
10 μL
200 μL
1250 μL
Tip Style:
Extended Length
Universal
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Tip Style
07-202-390 10 μL Extended Length
07-202-391 200 μL Universal
07-202-392 1250 μL Extended Length
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 07-202-390 Supplier Axygen™ Supplier No. MR10XT
Add to Cart
Add to Cart
This item is not returnable. View return policy

MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in Bulk, Racked, Filter and Reload packaging styles to meet the variety of needs in the lab setting.

The Axygen MultiRack tip system has been engineered to fit most of the pipettors in the market. It is an interchangeable complete system offered in bulk, racked, filter, and reload packaging styles to meet the variety of needs in the lab setting. The MultiRack tip system offers standard and extended length tips covering volumes from 1 μL to 1250 μL. For racked and reload packaging styles, the tips are pre-loaded into easy-load color-coded deck inserts. With the smaller footprint and reusable rack, this system uses 30% less plastic, minimizing the environmental impact. The stackable design saves critical bench space. Axygen MultiRack Tip System products are made in the USA and meet ISO standards. All tips are free of detectable RNase, DNase, DNA, and pyrogens. For more details visit our Certificates page at www.corning.com/lifesciences.
Features
• Reusable rack with reload systems 
• Smaller footprint and stackable design, saving valuable bench space
• Extended tips offered in 10, 300, and 1250 μL (1000 μL extended length) volumes
• Color-coded racks for easy volume identification

 

TRUSTED_SUSTAINABILITY

Specifications

Volume (Metric) 10 μL
Filtered Non-filtered
Sterility Non-sterile
Color Clear
Model MR-10XT
Certifications/Compliance ISO Standards
Clean Claims Free of detectable RNase, DNase, DNA, and Pyrogens
Tip Style Extended Length
Packaging Bulk
Quantity 1,000/pk, 10 pks/cs
Product Type Pipette Tips
Disposable Single-use
For Use With (Equipment) Universal Fit
Graduated Yes
Product Line MultiRack
Material Polyethylene
Show More Show Less
WARNING: Cancer and Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.