Learn More
Research Products International Corp MultiSample BioPulverizer with 12 Chambers and Pestles
SDP
Catalog No. 50550165
Encompass
Process up to 12 samples using the MultiSample Bio-Pulverizer. Cryogenic frozen tissue is turned into fine powder and ready for analysis. Place tissue into pre-chilled chamber, insert pestle and strike with a hammer. The compression force of the pestle instantly turns brittle tissue fine powder.
Manufactured of corrosion proof stainless steel for easy cleaning. Supplied with 12 pestles, 12 chamber wells and hammer.
Catalog No. 50-550-165 Supplier Research Products International Corp Supplier No. 141275
Process up to 12 samples using the MultiSample Bio-Pulverizer. Cryogenic frozen tissue is turned into fine powder and ready for analysis. Place tissue into pre-chilled chamber, insert pestle and strike with a hammer. The compression force of the pestle instantly turns brittle tissue fine powder.
Manufactured of corrosion proof stainless steel for easy cleaning. Supplied with 12 pestles, 12 chamber wells and hammer.