Learn More
Description
- MultiScreen™ 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms, including automated liquid handling, microplate readers, and other microplate compatible devices.
- ELISA microplates are available with medium or high binding capabilities.
Specifications
Specifications
| Barcode | No Barcode |
| Format | Strip Plate |
| Array | 8 x 12 |
| Color | Clear |
| Product Type | ELISA High Binding Plate |
| Material | Polystyrene |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Surface Treatment | Non-treated |
| Well Shape | Flat Bottom (F-well) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.