Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ PCRμ96, PCR96, and PCR384 Filter Plates
GSA_VA

Catalog No. p-155834 Shop All MilliporeSigma Products
Change view
Click to view available options
Volume (Metric) Well:
15 to 300 μL
150 μL
150 to 300 μL
No. of Wells:
96
Quantity:
10/Pk.
50/Pk.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Volume (Metric) Well No. of Wells Quantity
MSNU03010 150 to 300 μL 96 10/Pk.
LSKMPCR10 150 μL 96 10/Pk.
MSNU03050 15 to 300 μL 96 50/Pk.
LSKMPCR50 150 μL 96 50/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MSNU03010 Supplier MilliporeSigma™ Supplier No. MSNU03010
Only null left
Add to Cart
Add to Cart

Fully automatable PCR product purification

  • Nonsterile, clear polystyrene 96- and 384-well plates
  • Size-exclusion proprietary membranes retain DNA products at the surface
  • Complete in 10 to 12 minutes vacuum-based protocols without centrifugation, binding, or eluting
  • Suitable with 96- or 384-tip liquid handlers capable of pipetting ≤25μL/sec. or with plate shakers that have a 0.12 in. orbit at 1100rpm Working well volume, 10 to 300μL; nonrecoverable volume, <2μL
  • High-throughput format meets SBS (Society of Biomolecular Screening) standard
  • High-purity PCR product is ready for microarray spotting, sequencing, and cloning

Order Info

MultiScreen Vacuum Manifolds available

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance SBS
Format Standard
Array 8 x 12
Color Clear
Product Type MultiScreen PCRμ96
Material Styrene
Volume (Metric) Well 150 to 300 μL
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Purification of PCR products
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.