Learn More
Sigma Aldrich Fine Chemicals Biosciences MultiScreenR 8 well strip
Catalog No. 502908497
Encompass
MultiScreen(R) 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.
Catalog No. 50-290-8497 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M8CHNFX00
MultiScreen(R) 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.