Learn More
Sigma Aldrich Fine Chemicals Biosciences MultiScreenR 96-well Aplat
Catalog No. 502899994
Encompass
MultiScreen(R) 96-well polystyrene microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. Polystyrene microplates are appropriate for a variety of biochemical assays in addition to sample preparation and storage.
Catalog No. 50-289-9994 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSSWNFX40
MultiScreen(R) 96-well polystyrene microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. Polystyrene microplates are appropriate for a variety of biochemical assays in addition to sample preparation and storage.