Learn More
Sigma Aldrich Fine Chemicals Biosciences MultiScreenR 96-well ELISA
Catalog No. 502899989
Encompass
MultiScreen(R) 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.
Catalog No. 50-289-9989 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. MSEHNFX40
MultiScreen(R) 96-well ELISA microplates meet industry standard dimensions for use in common biochemical assay platforms including automated liquid handling microplate readers and other microplate compatible devices. ELISA microplates are available with medium or high binding capabilities.