Learn More
Beckman Coulter Multisizer™ 4e Smart Technology (ST) Footed Beakers
Catalog No. 15655128
Change view
Click to view available options
Capacity (Metric):
100 mL
Catalog No. Capacity (Metric)
15-655-128 100 mL
1 options
Catalog No. 15-655-128 Supplier Beckman Coulter Supplier No. A35595/DEL
Item Discontinued This item has been discontinued by the Fisher Scientific channel. Please to view product availability in your area.
Smart Technology Beakers fit the built-in beaker guide for proper positioning allowing the Multisizer 4e to deliver consistent, optimal results.
  • Unique design optimizes particle suspension
  • Beaker base is designed to fit a built-in guide on the sample platform 
  • Raised base of the beaker maximizes the volume of fluid the Analyzer is able to extract before drawing air through the aperture orifice
  • Combination of ST Beakers and the built-in guide allows the Multisizer 4e to deliver consistent, optimal results
TRUSTED_SUSTAINABILITY
Specifications
Product Type Footed Beaker
Capacity (Metric) 100 mL
Material Glass
For Use With (Equipment) Multisizer 4e Coulter Counter with Smart Technology (ST)