Learn More
THERMO SCI LSG 3P Thermo Fisher Scientific Multiskan FC, Filter-based, 96 and 384 Well, Shaker, 1 ea
Supplier:  THERMO SCI LSG 3P 51119000
Encompass_Preferred
The Thermo Scientific Multiskan FC Microplate Photometer offers reliable performance for absorbance measurements in various research and clinical applications. It is a filter-based instrument designed for professional personnel, measuring absorbance from suitable microplates and strips in 96-well plate formats, with internal shaking.
  • Wide wavelength range of 340 to 850 nm
  • Fast reading of 96-well plates
  • Large color screen and multiple language options for ease of use
  • Logical workflow with SkanIt Software for enhanced usability
  • USB port for data transfer and protocol import/export
  • Internal shaker with three selectable speeds
  • Built-in self-diagnostics and auto-calibration
  • Supports end point, dual wavelength, and kinetic assays
  • Internal memory stores 99 assay protocols and results
Catalog No. 50-950-417
Add to Cart
Request a Quote
Description
Description
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov