Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

THERMO SCI LSG 3P Thermo Fisher Scientific Multiskan FC Photometer, Incubator, 1 ea

Supplier:  THERMO SCI LSG 3P 51119100

Encompass_Preferred

The Multiskan FC with Incubator is a high-quality, filter-based microplate photometer designed for measuring absorbance from 96 and 384-well plate formats. It features integrated shaking and incubation capabilities for temperature-sensitive assays.

  • Measures absorbance from 96 and 384-well plates
  • Wavelength range of 340 to 850 nm
  • Incubator chamber with temperature range of 25°C to 50°C (max.) in 1°C increments, preventing condensation
  • Three linear shaking speeds: Slow, Medium, and Fast
  • Internal memory for 99 assay protocols and 100+ microplate assay results
  • One-channel optical system with tungsten halogen lamp and interference filters for wavelength selection

Catalog No. 50-950-418


May include imposed supplier surcharges.
Add to Cart
Request a Quote

Description

Description

TRUSTED_SUSTAINABILITY
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

WARNING: Cancer - www.P65Warnings.ca.gov
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.