Learn More
THERMO SCI LSG 3P Multiskan SkyHigh Microplate Spectrophotometer, Touchscreen
Supplier: THERMO SCI LSG 3P A51119700C
Thermo Scientific Multiskan SkyHigh Microplate Spectrophotometer is an easy to use instrument for photometric and turbidimetric applications, especially for DNA, RNA and protein analyses, including endpoint, kinetic, and spectral assays. Results are shown on Multiskan SkyHigh models with a large high-resolution touch screen. SkanIt Software is a powerful tool to address the challenging applications.
• Monochromator-based optical system for free selection of wavelengths from 200 nm to 1000 nm
• Compatible with 6-48, 96- and 384-well microplates or cuvettes with maximum plate height of 19.5 mm including possible lid
• Reads μDrop™ and μDrop Duo Plates for micro-volume DNA/RNA and protein analysis
• Separate optimized measurement modes for absorption and turbidimetric measurements
• Allows kinetic, spectral, and endpoint measurements for a variety of applications
• Fast operation: full spectrum of a well is done in less than 10 seconds