Learn More
Chemglass Life Sciences MultiStirrer, 6 Position, Digital, Magnetic, 100V, 50-60Hz, US Plug
SDP
Catalog No. 501531383
Encompass_Preferred
Chemglass 135mm round top ANALOG hot plate stirrer system. Hotplates feature many exclusive features and improved safety enhancements. The 135mm diameter heater top is scratchproof and chemically-resistant. The back corner of the housing has an M8 female thread to accept a vertical support rod. Note: Hot plates have been designed and manufactured in compliance with the following standards: CE, CSA and UL.
Catalog No. 50-153-1383 Supplier Chemglass Life Sciences Supplier No. CG1998V06
Chemglass 135mm round top ANALOG hot plate stirrer system. Hotplates feature many exclusive features and improved safety enhancements. The 135mm diameter heater top is scratchproof and chemically-resistant. The back corner of the housing has an M8 female thread to accept a vertical support rod. Note: Hot plates have been designed and manufactured in compliance with the following standards: CE, CSA and UL.