Learn More
Chemglass Life Sciences MultiStirrer, 6 Position, Digital, Magnetic, 240V, 50-60Hz
SDP
Catalog No. 501531384
Encompass_Preferred
MultiStirrer, 6 Position, Digital, Magnetic, 240V, 50-60Hz. Multiposition stirrer with Stainless Steel top. Multistirrer Digital is the only multi-position magnetic stirrer with a timer, giving the ability to program the unit for unattended operation. Stirrer has a bright digital display to accurately set the stirring speed. The speed is adjustable in 10 rpm increments from 80 up to 1500 rpm. SpeedServo ensures constant speed even when the viscosity changes. *ITEM HAS A EUROPEAN PLUG FOR USE OVERSEAS. CG-1998-V06 which is for use in the U.S*
Catalog No. 50-153-1384 Supplier Chemglass Life Sciences Supplier No. CG1998V06E
MultiStirrer, 6 Position, Digital, Magnetic, 240V, 50-60Hz. Multiposition stirrer with Stainless Steel top. Multistirrer Digital is the only multi-position magnetic stirrer with a timer, giving the ability to program the unit for unattended operation. Stirrer has a bright digital display to accurately set the stirring speed. The speed is adjustable in 10 rpm increments from 80 up to 1500 rpm. SpeedServo ensures constant speed even when the viscosity changes. *ITEM HAS A EUROPEAN PLUG FOR USE OVERSEAS. CG-1998-V06 which is for use in the U.S*