Learn More
Chemglass Life Sciences MultiStirrer, 15 Position, Digital, Magnetic, 100V, 50-60Hz, US Plug
SDP
Catalog No. 501531385
Encompass_Preferred
Multiposition stirrer with stainless steel top. Multistirrer Digital is the only multi-position magnetic stirrer with a timer, giving the ability to program the unit for unattended operation. Stirrer has a bright digital display to accurately set the stirring speed. The speed is adjustable in 10 rpm increments from 80 up to 1500 rpm. SpeedServo ensures constant speed even when the viscosity changes.
Catalog No. 50-153-1385 Supplier Chemglass Life Sciences Supplier No. CG1998V15
Multiposition stirrer with stainless steel top. Multistirrer Digital is the only multi-position magnetic stirrer with a timer, giving the ability to program the unit for unattended operation. Stirrer has a bright digital display to accurately set the stirring speed. The speed is adjustable in 10 rpm increments from 80 up to 1500 rpm. SpeedServo ensures constant speed even when the viscosity changes.