Learn More
Siemens Healthineers Multistix™ and Uristix™ Reagent Strips
Catalog No. 23042351 Shop All Siemens Healthineers Products
Change view
Click to view available options
Detectable Analytes:
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen
Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein
Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity
Glucose, Leukocyte, Nitrite, Protein
Description:
Multistix 10SG
Multistix 5
Multistix 7
Multistix 8 SG
Uristix 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Detectable Analytes Description
23-042-351 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen Multistix 5
23-042305 Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity, Urobilinogen Multistix 10SG
AM-2164 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Specific Gravity Multistix 8 SG
AM-2166 Glucose, Leukocyte, Nitrite, Protein Uristix 4
23-042310 Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein Multistix 7
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Catalog No. 23-042-351 Supplier Siemens Healthineers Supplier No. 10337415
Add to Cart
Edge
Add to Cart
This item is not returnable. View return policy
Visually and instrumentally readable urinalysis strips
  • Includes testing for a variety of parameters
  • Supplied in lightweight, unbreakable, plastic vials
  • Provide diagnostically useful information about the status of carbohydrate metabolism, kidney and liver function, acid-base balance and bacteriuria-pyuria
TRUSTED_SUSTAINABILITY
Specifications
Description Multistix 5
Detectable Analytes Bilirubin, Blood, Glucose, Ketone, Leukocyte, Nitrite, pH, Protein, Urobilinogen
For Use With (Application) Urinalysis
Quantity 100 Pack