Learn More
Sigma Aldrich Fine Chemicals Biosciences MultiThermTM shaker with h1EA
Catalog No. 502908971
Encompass
The MultiTherm shaker is a temperature controlled vortexer useful for a variety of molecular biology applications. Despite its modest 8 x 11 in. footprint all popular tubes and plates are compatible including as many as 6 x 50 mL tubes.Speed time and temperature settings are continuously visible on the LCD simultaneously showing both actual and selected values. Integral over-temperature control ensures long life safety and sample integrity.Blocks available separately accomodate tubes from 0.2 mL to 50 mL.Heating rate 5C/min.Cooling rate 2.5C/min.Speed increment 10 rpmTimer 1min. to 99 hrs 99 min./continuousContoured wells provide uniform heating
Catalog No. 50-290-8971 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. Z7557531EA
The MultiTherm shaker is a temperature controlled vortexer useful for a variety of molecular biology applications. Despite its modest 8 x 11 in. footprint all popular tubes and plates are compatible including as many as 6 x 50 mL tubes.Speed time and temperature settings are continuously visible on the LCD simultaneously showing both actual and selected values. Integral over-temperature control ensures long life safety and sample integrity.Blocks available separately accomodate tubes from 0.2 mL to 50 mL.Heating rate 5C/min.Cooling rate 2.5C/min.Speed increment 10 rpmTimer 1min. to 99 hrs 99 min./continuousContoured wells provide uniform heating