Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MEGAComfort MultiThotic™ 3-in-1 Orthotics, Dual Layer Memory Foam Insole

Catalog No. 18813512
Change view
Click to view available options
Size:
Men's: 6-7, Women's: 8-9
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-813-512 Men's: 6-7, Women's: 8-9
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 18-813-512 Supplier MEGAComfort Supplier No. MTM67/W89
This item is not returnable. View return policy

Orthotic insole with medical-grade polyplastic designed by podiatrists, Dual layer 100% memory foam, Clinically proven field tested to reduce pain and fatigue, Shock absorption comfort

  • Uses patented combination of high-density dual layer 100% memory foam plus two re-attachable orthotic arch supports (medical grade materials)
  • Light green flexible arch support recommended for medium/regular arch type
  • Dark green rigid arch support recommended for low arch/pronation support
  • Provides adaptable comfort and shock absorption while simultaneously allowing one to vary the degree of heel, arch support and motion control
  • Ideal for stationary, mobile and working outdoors
  • For use with industrial (safety and steel-toe footwear), casual and work footwear
  • Soft contoured arch support for balance, comfort and cushioning
  • Silver ion anti-microbial technology in top cloth offering anti-bacterial, anti- perspiration and anti-odor
  • Machine washable

Specifications

Color Black, Blue, Yellow
For Use With (Application) For stationary, mobile and working outdoors
For Use With (Equipment) Industrial (safety and steel-toe), casual and work footwear
Material Memory Foam, Plastic
Size Men's: 6-7, Women's: 8-9
Gender Unisex
Waterproof No
Product Type Orthotic insole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.