Learn More
Braintree Scientific Multivette 500 - 500 ul,Lithium-Heparin Gel, 1,000/cs
SDP
Catalog No. 501955310
Encompass
Microvette 500 Blood collection with collection rim The new Microvette tubes are ideal for the collection of blood using the gravity-flow principle of collection. Any part of the entire rim can be used. The Microvette offers all the advantages of a modern blood collection system: Special twist cap design is easy to open and minimizes aerosol effect. Cylindrical outer container is ideal for barcode or patient labels **Sold for research purposes only and not for human use**
Catalog No. 50-195-5310 Supplier Braintree Scientific Supplier No. MV500201346
Microvette 500 Blood collection with collection rim The new Microvette tubes are ideal for the collection of blood using the gravity-flow principle of collection. Any part of the entire rim can be used. The Microvette offers all the advantages of a modern blood collection system: Special twist cap design is easy to open and minimizes aerosol effect. Cylindrical outer container is ideal for barcode or patient labels **Sold for research purposes only and not for human use**