Learn More
Braintree Scientific Multivette 600 - 600 ul, Serum/Clotting Activator, 100/bx
SDP
Catalog No. 501955313
Encompass
Multivette 600 Capillary and venous blood collection in one system. The unique Multivette 600 is the only product that can collect both venous and capillary blood. For venous blood collection, a Luer needle is connected to the capillary tube of the Multivette, which automatically fills by normal venous pressure. Capillary blood is performed using the end-to-end principle. The special pre-assembled capillary tube is designed to hold 600?L of blood. Depending on individual requirements, the Microvette is available with a cylindrical inner tube for optimum mixing results or a conical inner tube for a high serum/plasma supernatant. Cylindrical outer container is ideal for barcodes and patient labels. Special cap design minimizes aerosol effect when tube is opened and provides safe sealing features. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5313 Supplier Braintree Scientific Supplier No. MV600151670BX
Multivette 600 Capillary and venous blood collection in one system. The unique Multivette 600 is the only product that can collect both venous and capillary blood. For venous blood collection, a Luer needle is connected to the capillary tube of the Multivette, which automatically fills by normal venous pressure. Capillary blood is performed using the end-to-end principle. The special pre-assembled capillary tube is designed to hold 600?L of blood. Depending on individual requirements, the Microvette is available with a cylindrical inner tube for optimum mixing results or a conical inner tube for a high serum/plasma supernatant. Cylindrical outer container is ideal for barcodes and patient labels. Special cap design minimizes aerosol effect when tube is opened and provides safe sealing features. **Sold for research purposes only and not for human use**