Learn More
Molecular Devices Molecular Devices Microplate Reader, 6-1536-Well, Dual PMT, User-Upgradeable
Catalog No. 5000014675
Encompass_Preferred
The SpectraMax Paradigm multi-mode microplate reader from Molecular Devices is a user-upgradeable detection platform for absorbance, fluorescence, time-resolved fluorescence, fluorescence polarization, and luminescence assays. It supports 6- to 1536-well plates and modular detection cartridges for configurable optics.
  • User-upgradeable detection cartridges for configurable optics and modes.
  • Supports 6- to 1536-well plate formats.
  • Dual photomultiplier tubes for enhanced sensitivity and throughput.
  • Wide absorbance range 230-1000 nm and fluorescence excitation/emission coverage.
  • Integrated temperature control, plate shaking, and robotic compatibility.
  • High dynamic range greater than six decades for quantitative assays.
  • Compact footprint: 39 cm W x 45 cm H x 63 cm D.
Catalog No. 50-000-14675 Supplier Molecular Devices Supplier No. PARADIGM
Add to Cart
Add to Cart
Request a Quote
The SpectraMax Paradigm multi-mode microplate reader from Molecular Devices is a user-upgradeable detection platform for absorbance, fluorescence, time-resolved fluorescence, fluorescence polarization, and luminescence assays. It supports 6- to 1536-well plates and modular detection cartridges for configurable optics.
  • User-upgradeable detection cartridges for configurable optics and modes.
  • Supports 6- to 1536-well plate formats.
  • Dual photomultiplier tubes for enhanced sensitivity and throughput.
  • Wide absorbance range 230-1000 nm and fluorescence excitation/emission coverage.
  • Integrated temperature control, plate shaking, and robotic compatibility.
  • High dynamic range greater than six decades for quantitative assays.
  • Compact footprint: 39 cm W x 45 cm H x 63 cm D.