Learn More
Anton Paar Multiwave GO Plus microwave digester combines single- and multi-mode benefits via Directed Multimode Cavity (DMC). Digests up to 12 samples with fast heating/cooling and high throughput. Single-Vessel Mode enables optimization, while SmartVent ensures safe, easy handling.