Learn More
Anton Paar Multiwave GO Plus microwave digester combines single- and multi-mode benefits via Directed Multimode Cavity (DMC). Digests up to 12 samples with fast heating/cooling and high throughput. Single-Vessel Mode enables optimization, while SmartVent ensures safe, easy handling.
Catalog No. 5000580519
Encompass_Preferred
SpecificationsNumber of Vessels: 12Volume: 50 mLMaterial: PTFE-TFMHF resistanceTemperature control: IR temperature sensorPressure control: SmartVent technologyApplications: Routine Sample - Biological and environmental samples, EPA procedures, food, feed, aqueous samples
Catalog No. 50-005-80519 Supplier Anton Paar Supplier No. 231708
Please contact your sales representative for a quote or further ordering instructions.
SpecificationsNumber of Vessels: 12Volume: 50 mLMaterial: PTFE-TFMHF resistanceTemperature control: IR temperature sensorPressure control: SmartVent technologyApplications: Routine Sample - Biological and environmental samples, EPA procedures, food, feed, aqueous samples