Learn More
Chemglass Life Sciences Reservoir, Multi-Well, 8-Channel, Non-Sterile, 10/Pk, 50/Case
SDP
Catalog No. 501897722
Encompass_Preferred
Nest Robotic Reagent Reservoirs are polypropylene reservoirs that are designed for use with automated and robotic systems. • Low profile design for automation systems/robots • Temperature range from -80 °C to +121 ° • Low-retention for reducing sample loss • Made of Clear Polypropylene • Compatible with DMSO and inert to water • Conforms to ANSI/SBS Standards
Catalog No. 50-189-7722 Supplier Chemglass Life Sciences Supplier No. CGN3797HP8
Add to Cart
Add to Cart
Nest Robotic Reagent Reservoirs are polypropylene reservoirs that are designed for use with automated and robotic systems. • Low profile design for automation systems/robots • Temperature range from -80 °C to +121 ° • Low-retention for reducing sample loss • Made of Clear Polypropylene • Compatible with DMSO and inert to water • Conforms to ANSI/SBS Standards