Learn More
Chemglass Life Sciences Reservoir, Multi-Well, 96-Channel, Non-Sterile, 10/Pk, 50/Case
SDP
Catalog No. 501897724
Encompass_Preferred
Nest Robotic Reagent Reservoirs are polypropylene reservoirs that are designed for use with automated and robotic systems. • Low profile design for automation systems/robots • Temperature range from -80 °C to +121 ° • Low-retention for reducing sample loss • Made of Clear Polypropylene • Compatible with DMSO and inert to water • Conforms to ANSI/SBS Standards
Catalog No. 50-189-7724 Supplier Chemglass Life Sciences Supplier No. CGN3797HP96
Nest Robotic Reagent Reservoirs are polypropylene reservoirs that are designed for use with automated and robotic systems. • Low profile design for automation systems/robots • Temperature range from -80 °C to +121 ° • Low-retention for reducing sample loss • Made of Clear Polypropylene • Compatible with DMSO and inert to water • Conforms to ANSI/SBS Standards