Learn More
US Biomax Inc Multiple organ normal tissue array, 44 cases/48 cores, replacing BN00011a
SDP
Catalog No. NC3244994 Shop All US Biomax Inc Products
Encompass
Multiple organ normal tissue microarray, containing 8 types of normal human organs (esophagus, stomach, liver, colon, rectum, lung, kidney, breast), each type taken from 6 normal human individuals, single core per block
Catalog No. NC3244994 Supplier US Biomax Inc Supplier No. BN00011B
Multiple organ normal tissue microarray, containing 8 types of normal human organs (esophagus, stomach, liver, colon, rectum, lung, kidney, breast), each type taken from 6 normal human individuals, single core per block