Learn More
Cell Biolabs Inc QuickTiter™ MuLV Core Antigen ELISA Kit, 96 assays
Supplier:  Cell Biolabs Inc VPK156
Encompass_Preferred
Cell Biolabs' QuickTiter™ MuLV Core Antigen ELISA Kit measures the p30 core antigen of MuLV with sensitivity as low as 300 pg/mL. The kit provides sufficient reagents for 96 assays including standards and unknown samples.
Catalog No. NC0394716