Learn More
Eurofins Discoverx muMAP Kinase 2/Erk2, unactive, 50 µg. (14-198)
Supplier:  Eurofins Discoverx 14198
Encompass
N-terminal GST-tagged recombinant, murine full length p42 MAP kinase 2/Erk2. For detailed product information, visit www.discoverx.com/proteins.
Catalog No. 50-199-8723