Learn More
New England Biolabs, Inc. Mung Bean Nuclease – 7500 units
SDP
Catalog No. 50811664
Encompass
Mung Bean Nuclease is a single-strand specific DNA and RNA endonuclease which will degrade single-stranded extensions leaving blunt, ligatable ends.
Catalog No. 50-811-664 Supplier New England Biolabs, Inc. Supplier No. M0250L
Mung Bean Nuclease is a single-strand specific DNA and RNA endonuclease which will degrade single-stranded extensions leaving blunt, ligatable ends.