Learn More
Abcam MUP disodium salt trihydrate, Fluorogenic phosphatase substrate, 1G
Catalog No. 502704602
Encompass_Preferred
MW 354.16 g/mol. Suitable as a fluorogenic substrate for phosphatases. Fluorescence: λex 319 nm; λem 384 nm (pH 9.0); λex 360 nm; λem 449 nm (Reaction product).
The product is subject to the following: Abcam Restricted Use Statement
Catalog No. 50-270-4602 Supplier Abcam Supplier No. AB1465211G
MW 354.16 g/mol. Suitable as a fluorogenic substrate for phosphatases. Fluorescence: λex 319 nm; λem 384 nm (pH 9.0); λex 360 nm; λem 449 nm (Reaction product).
The product is subject to the following: Abcam Restricted Use Statement