Learn More
Apexbio Technology LLC Mupirocin 12650-69-0 10mM (in 1mL DMSO)
SDP
Catalog No. 5000062880
Encompass_Preferred
Mupirocin is an antibiotic targeting bacterial protein synthesis by inhibition of isoleucyl-tRNA synthetase Structurally analogous to isoleucine it binds the enzyme s active site blocking formation of isoleucyl-tRNA thus arresting bacterial protein production with reported MIC values around 0 05 g/ml for Staphylococcus aureus Primarily active against Gram-positive strains notably methicillin-resistant Staphylococcus aureus (MRSA) it is widely applied in biomedical research focused on antimicrobial resistance soft tissue infections biofilm studies and nasal colonization control strategies
Catalog No. 50-000-62880 Supplier Apexbio Technology LLC Supplier No. B48725.1
Mupirocin is an antibiotic targeting bacterial protein synthesis by inhibition of isoleucyl-tRNA synthetase Structurally analogous to isoleucine it binds the enzyme s active site blocking formation of isoleucyl-tRNA thus arresting bacterial protein production with reported MIC values around 0 05 g/ml for Staphylococcus aureus Primarily active against Gram-positive strains notably methicillin-resistant Staphylococcus aureus (MRSA) it is widely applied in biomedical research focused on antimicrobial resistance soft tissue infections biofilm studies and nasal colonization control strategies