Learn More
Selleck Chemical LLC Mupirocin-25mg
SDP
Supplier:  Selleck Chemical LLC S429725MG
Encompass_Preferred
Mupirocin(BRL-4910A,Pseudomonic acid) is an isoleucyl t-RNA synthetase inhibitor, used in the treatment of bacterial skin infections.
Catalog No. 50-239-0044