Learn More
TARGETMOL CHEMICALS INC MUPIROCIN 50MG
SDP
Catalog No. 502577379
Encompass_Preferred
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mupirocin (BRL-4910A) is a RNA synthetase inhibitor antibacterial. It has shown excellent activity against gram-positive staphylococci and streptococci. purity: 98%
Catalog No. 50-257-7379 Supplier TARGETMOL CHEMICALS INC Supplier No. T146550MG
Also available in 5 mg 10 mg 25 mg 100 mg 200 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. Mupirocin (BRL-4910A) is a RNA synthetase inhibitor antibacterial. It has shown excellent activity against gram-positive staphylococci and streptococci. purity: 98%