Learn More
Selleck Chemical LLC Mupirocin calcium S5070-25mg
SDP
Catalog No. 503111961
Encompass_Preferred
Mupirocin Calcium (Pseudomonic acid A) is the calcium salt form of mupirocin a natural crotonic acid derivative extracted from Pseudomonas fluorescens which inhibits bacterial protein synthesis and is used as an antibiotic
Catalog No. 50-311-1961 Supplier Selleck Chemical LLC Supplier No. S507025MG
Mupirocin Calcium (Pseudomonic acid A) is the calcium salt form of mupirocin a natural crotonic acid derivative extracted from Pseudomonas fluorescens which inhibits bacterial protein synthesis and is used as an antibiotic