Learn More
Apexbio Technology LLC MUPIROCIN-CALCIUM-50MG
SDP
Catalog No. 5000568832
Encompass_Preferred
Mupirocin calcium (CAS No 115074-43-6) is an antibiotic compound used in biomedical research primarily in the study of bacterial infections Originating from Pseudomonas fluorescens its mechanism of action involves inhibiting bacterial protein synthesis by binding to isoleucyl-tRNA synthetase disrupting the formation of isoleucyl-tRNA This results in effective antibacterial activity typically observed in the nanomolar to low micromolar range in vitro Mupirocin calcium is commonly used in cell models and animal studies to investigate its efficacy and mechanism against Gram-positive bacteria particularly methicillin-resistant Staphylococcus aureus (MRSA) It serves as a valuable tool in drug screening for developing new antibacterial agents Experimental concentrations typically depend on the specific research design
Catalog No. 50-005-68832 Supplier Apexbio Technology LLC Supplier No. C650050
Mupirocin calcium (CAS No 115074-43-6) is an antibiotic compound used in biomedical research primarily in the study of bacterial infections Originating from Pseudomonas fluorescens its mechanism of action involves inhibiting bacterial protein synthesis by binding to isoleucyl-tRNA synthetase disrupting the formation of isoleucyl-tRNA This results in effective antibacterial activity typically observed in the nanomolar to low micromolar range in vitro Mupirocin calcium is commonly used in cell models and animal studies to investigate its efficacy and mechanism against Gram-positive bacteria particularly methicillin-resistant Staphylococcus aureus (MRSA) It serves as a valuable tool in drug screening for developing new antibacterial agents Experimental concentrations typically depend on the specific research design