Learn More
Medchemexpress LLC Mupirocin lithium 25mg
SDP
Catalog No. 5000205904
Encompass_Preferred
Mupirocin lithium is an antibiotic Mupirocin lithium inhibits bacterial isoleucyl-tRNA synthetase blocking protein synthesis Mupirocin lithium has high activity against Gram-positive bacteria such as Staphylococcus and Streptococcus as well as some Gram-negative bacteria (such as Haemophilus influenzae) Mupirocin lithium can be used in the research of diseases such as skin infections (such as MRSA infections) and chronic sinusitis[1][2][3][4][5][6]
Catalog No. 50-002-05904 Supplier Medchemexpress LLC Supplier No. 25MGHYW108875
Mupirocin lithium is an antibiotic Mupirocin lithium inhibits bacterial isoleucyl-tRNA synthetase blocking protein synthesis Mupirocin lithium has high activity against Gram-positive bacteria such as Staphylococcus and Streptococcus as well as some Gram-negative bacteria (such as Haemophilus influenzae) Mupirocin lithium can be used in the research of diseases such as skin infections (such as MRSA infections) and chronic sinusitis[1][2][3][4][5][6]