Learn More
TARGETMOL CHEMICALS INC MURAGLITAZAR 5MG
SDP
Catalog No. 502581020
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Muraglitazar (BMS-298585) is a PPAR (alpha)/(gamma) dual agonist for the treatment of type 2 diabetes and associated dyslipidemia. Muraglitazar shows potent activity in vitro at human PPAR(alpha) and PPAR(gamma) with EC50 of 320 nM and 110 nM respectively. purity: 99%
Catalog No. 50-258-1020 Supplier TARGETMOL CHEMICALS INC Supplier No. T215875MG
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Muraglitazar (BMS-298585) is a PPAR (alpha)/(gamma) dual agonist for the treatment of type 2 diabetes and associated dyslipidemia. Muraglitazar shows potent activity in vitro at human PPAR(alpha) and PPAR(gamma) with EC50 of 320 nM and 110 nM respectively. purity: 99%