Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Murakami Reagent for Etching Steel, Reagents

Catalog No. 01334704
Change view
Click to view available options
Quantity:
500 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-704 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 01-334-704 Supplier Reagents Holdings Llc Supplier No. CM140000500A
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Murakami Reagent for Etching Steel, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Chemical Identifiers

CAS 7732-18-5 , 13746-66-2
Molecular Formula H2O
Molecular Weight (g/mol) 18.02
InChI Key XLYOFNOQVPJJNP-UHFFFAOYSA-N
IUPAC Name water
SMILES O

Specifications

Color Yellow
Grade Reagent
Packaging Poly Bottle
Specific Gravity 1.0891
pH >13
Quantity 500 mL
Sterility Non-sterile
Physical Form Liquid
Chemical Name or Material Murakami Reagent for Etching Steel
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.