Learn More
Medchemexpress LLC HY-127090 5mg , Muramyl dipeptide CAS:53678-77-6 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1270905MG
Encompass_Preferred
HY-127090 5mg Muramyl dipeptide CAS: 53678-77-6 Muramyl dipeptide (MDP) is a synthetic immunoreactive peptide, consisting of N-acetyl muramic acid attached to a short amino acid chain of L-Ala-D-isoGln. Muramyl dipeptide is an inducer of bone formation through induction of Runx2. Muramyl dipeptide directly enhances osteoblast differentiation by up-regulating Runx2 gene expression through MAPK pathways. Muramyl dipeptide is a NLRP1 agonist. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-226-1731