Learn More
Medchemexpress LLC Muramyl dipeptide (MDP) | 53678-77-6 | 99.1% | 10 MG
SDP
Catalog No. 5000341562
Encompass_Preferred
Muramyl dipeptide (MDP) is a synthetic immunoreactive peptide composed of N-acetyl muramic acid attached to an L-Ala-D-isoGln amino acid chain. It acts as an inducer of bone formation by promoting Runx2 and directly enhances osteoblast differentiation via MAPK pathways. MDP is also a NLRP1 agonist.
  • Synthetic immunoreactive peptide
  • Induces bone formation through induction of Runx2
  • Directly enhances osteoblast differentiation via MAPK pathways
  • Acts as a NLRP1 agonist
  • Increases protein expression of Runx2 in a dose-dependent manner
  • Increases mRNA levels of Runx2 in a dose-dependent manner
  • Indirectly attenuates osteoclast differentiation
  • Alleviates bone loss induced by osteoporosis in animal models
  • Significantly enhances trabecular bone volume and number in vivo
Catalog No. 50-003-41562 Supplier Medchemexpress LLC Supplier No. HY12709010MG
Muramyl dipeptide (MDP) is a synthetic immunoreactive peptide composed of N-acetyl muramic acid attached to an L-Ala-D-isoGln amino acid chain. It acts as an inducer of bone formation by promoting Runx2 and directly enhances osteoblast differentiation via MAPK pathways. MDP is also a NLRP1 agonist.
  • Synthetic immunoreactive peptide
  • Induces bone formation through induction of Runx2
  • Directly enhances osteoblast differentiation via MAPK pathways
  • Acts as a NLRP1 agonist
  • Increases protein expression of Runx2 in a dose-dependent manner
  • Increases mRNA levels of Runx2 in a dose-dependent manner
  • Indirectly attenuates osteoclast differentiation
  • Alleviates bone loss induced by osteoporosis in animal models
  • Significantly enhances trabecular bone volume and number in vivo