Learn More
Sigma Aldrich Fine Chemicals Biosciences Murashige and Skoog Vitamin Solution, 50 mL
Catalog No. NC1137540
Encompass
1000 ×, liquid, plant cell culture tested, color: colorless, pH: 5.5-7.5, storage temp: 2-8°C.
Catalog No. NC1137540 Supplier Sigma Aldrich Fine Chemicals Biosciences Supplier No. M390050ML
1000 ×, liquid, plant cell culture tested, color: colorless, pH: 5.5-7.5, storage temp: 2-8°C.