Learn More
PHYTOTECHNOLOGY LABS Murashige & Skoog Modified Multiplication Medium, 1 L
Catalog No. NC1702448
Encompass_Preferred
This medium may be sold as Murashige and Skoog Shoot Multiplication Medium C by some media suppliers.Contains the macro- and micronutrients as described by Murashige and Skoog (1962) and the vitamins described by Linsmaier and Skoog (1965). Also contains (mg/L): 148 Sodium Phosphate Monobasic, 80 Adenine Hemisulfate, 1.0 Kinetin, 0.1 NAA, and 30,000 Sucrose.
Catalog No. NC1702448 Supplier PHYTOTECHNOLOGY LABS Supplier No. M5551L
This medium may be sold as Murashige and Skoog Shoot Multiplication Medium C by some media suppliers.Contains the macro- and micronutrients as described by Murashige and Skoog (1962) and the vitamins described by Linsmaier and Skoog (1965). Also contains (mg/L): 148 Sodium Phosphate Monobasic, 80 Adenine Hemisulfate, 1.0 Kinetin, 0.1 NAA, and 30,000 Sucrose.