Learn More
Bioworld Murashige African Violet/Gloxinia Multiplication Medium, 50 L
Catalog No. 50254857
Encompass_Preferred
Murashige African Violet/Gloxinia Multiplication MediumContains Macro and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) modified with addition of Sodium Phosphate Monobasic, IAA, and KinetinSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.66 g/LpH: 3.5–4.5Soluble in water
Catalog No. 50-254-857 Supplier Bioworld Supplier No. 306301474
Murashige African Violet/Gloxinia Multiplication MediumContains Macro and Micronutrients as described by Murashige & Skoog (1962) and Vitamins as described by Linsmaier and Skoog (1965) modified with addition of Sodium Phosphate Monobasic, IAA, and KinetinSucrose, Cytokines, and Auxins should be added as needed Provided as 1 unit to make 10L, 25L, or 50L of medium, or as 10 units to make 1L of mediumPlant tissue culture testedWorking Concentration: 4.66 g/LpH: 3.5–4.5Soluble in water