Learn More
Sigma Aldrich Fine Chemicals Biosciences Murashige and Skoog Basal
Supplier:  Sigma Aldrich Fine Chemicals Biosciences M551950L
Encompass
Murashige and Skoog medium is a widely used plant tissue culture growth medium. MS Basal Medium contains macronutrients that include high levels of nitrate and organic additives such as agar sugars vitamins and growth regulators. Important growth regulators frequently added to MS include IAA (auxin/morphogen) and the Kinetin (cytokinin/cell division promoter).
Catalog No. 50-290-0472